Skip to Content
Merck
All Photos(1)

Key Documents

791281P

Avanti

α-mycolic acid, methoxy cis

powder

Synonym(s):

Cyclopropaneeicosanoic acid, β-hydroxy-2-[(17S,18S)-17-methoxy-18-methylhexatriacontyl]-α-tetracosyl-, (αR,βR,1R,2S)-

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$860.00

$860.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$860.00

About This Item

Empirical Formula (Hill Notation):
C85H168O4
CAS Number:
Molecular Weight:
1254.24
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$860.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (791281P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791281P

lipid type

bacterial lipids

shipped in

dry ice

storage temp.

−20°C

General description

α-mycolic acid (MA), methoxy cis comprises 84–88 carbon atoms like the other types keto and hydroxy MA.[1] All the MA types are synthesized from hydroxylated intermediate, with methoxy M generated by the action of enzyme methoxylase.[1]
Mycolic acid is a long Fatty acid found in the cell walls of the mycolata taxon, a group of bacteria that includes Mycobacterium tuberculosis, the causative agent of the disease tuberculosis.

Biochem/physiol Actions

Methoxy mycolic acids are antigenic and are specific for pathogenic mycobacterial class.[2] The three types of mycolic acids including the methoxy-MA display variation in their antigenicity with reference to antibody recognition.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (791281P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hedia Marrakchi et al.
Chemistry & biology, 21(1), 67-85 (2014-01-01)
Mycolic acids are major and specific lipid components of the mycobacterial cell envelope and are essential for the survival of members of the genus Mycobacterium that contains the causative agents of both tuberculosis and leprosy. In the alarming context of
Nabil Tahiri et al.
Angewandte Chemie (International ed. in English), 59(19), 7555-7560 (2020-02-19)
In Mycobacterium tuberculosis, mycolic acids and their glycerol, glucose, and trehalose esters ("cord factor") form the main part of the mycomembrane. Despite their first isolation almost a century ago, full stereochemical evaluation is lacking, as is a scalable synthesis required
Heena Ranchod et al.
PloS one, 13(8), e0200298-e0200298 (2018-08-10)
Mycolic acids (MA) are major, species-specific lipid components of Mycobacteria and related genera. In Mycobacterium tuberculosis, it is made up of alpha-, methoxy- and keto-MA, each with specific biological functions and conformational characteristics. Antibodies in tuberculosis (TB) patient sera respond

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service