Skip to Content
Merck
All Photos(1)

Key Documents

790326P

Avanti

Fluorescent Transfection Reagent

Avanti Research - A Croda Brand 790326P, powder

Synonym(s):

Fluorescent Transfection Reagent, Fluorescent Transfection Reagent, contains DOTAP:DOPE (1:1, w/w) + 0.1 wt.% NBD-DOPE

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$758.00

$758.00


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$758.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.83

$758.00


Estimated to ship on01 April 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (790326P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790326P

technique(s)

transfection: suitable

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescent transfection reagent has the cationic lipid 1,2-dioleoyloxypropyl)-N,N,N-trimethylammonium chloride (DOTAP) and the neutral lipid DOPE (1,2-dioleoyl-sn-glycero-3-phosphoethanolamine in the ratio 1:1 w/w along with 0.1 wt% N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)dioleoyl (NBD) DOPE.[1] NBD-DOPE is a fluorescence probe [2] with NBD attached to headgroup of DOPE.[3]

Biochem/physiol Actions

The DOTAP:DOPE with 0.1% NBD-DOPE liposome (LipNBD) reconstituted with transferrin called the transferrin liposome-nitrobenzoxadiazole (Tf-LipNBD) is useful as a probe. It provides improved image contrast in in vitro magnetic resonance imaging (MRI) of tumor cells.[4] Fluorescent transfection reagent aids in understanding fibroplex membrane structure[2] and fibroin interaction with cationic lipids.[1] N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl)dioleoyl (NBD) is used in model membranes studies and is useful in visualizing membrane domains.[3]

Packaging

5 mL Clear Glass Sealed Ampule (790326P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The influence of NBD fluorescent probe on model membranes containing POPC and DPPC
Weng CJ, et al.
Molecular Membrane Biology, 33(1-2), 23-28 (2016)
Dual probe with fluorescent and magnetic properties for imaging solid tumor xenografts
Shan L, et al.
Molecular Imaging, 6(2), 7290-2007 (2007)
Efficiency data of intracellular recombinant protein delivery using cationic lipid coated silk fibroin particle
Kim WJ, et al.
Data in Brief, 13, 683-691 (2017)
Woo-Jin Kim et al.
Biomaterials, 122, 154-162 (2017-01-26)
Directly delivering therapeutic proteins into cells has promise as an intervention without side effects for protein deficiencies caused by genetic defects. However, as negatively charged macromolecules, proteins require carriers for achieving cellular uptake and maintaining their activity in the cytoplasm.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service