Skip to Content
Merck
All Photos(1)

Key Documents

700167P

Avanti

18:1 stigmasteryl glucose

6-O-oleoyl-stigmasteryl-β-D-glucose, powder

Synonym(s):

3-O-[(6′-O-oleoyl)-β-D-glucopyranosyl] stigmasterol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$288.00

$288.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$288.00

About This Item

Empirical Formula (Hill Notation):
C53H90O7
CAS Number:
Molecular Weight:
839.28
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$288.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700167P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700167P

shipped in

dry ice

storage temp.

−20°C

General description

18:1 stigmasteryl glucose comprises 18:0 (oleic) acyl group at the C6 position in the glucose molecule and is an acyl steryl glucoside (ASG).[1] 18:1 (oleic) is a major ASG in Arabidopsis.[2]

Biochem/physiol Actions

Mutation in UDP-glucose:sterol glucosyltransferase (UGT) enzymes deplete the 18:1 (oleic) and other acyl steryl glucoside (ASG) levels.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700167P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathrin Schrick et al.
Lipids, 47(2), 185-193 (2011-08-11)
Establishment of sensitive methods for the detection of cellular sterols and their derivatives is a critical step in developing comprehensive lipidomics technology. We demonstrate that electrospray ionization tandem (triple quadrupole) mass spectrometry (ESI-MS/MS) is an efficient method for monitoring steryl
Daniel F Stucky et al.
Journal of experimental botany, 66(1), 189-201 (2014-10-16)
Steryl glucosides (SG) are abundant steroid conjugates in plant membranes. Beyond structural roles in lipid bilayers, functions in sugar transport, storage, and/or signalling are predicted. UDP-glucose:sterol glucosyltransferase 80A2 (UGT80A2) and UGT80B1, which share similarity to fungal counterparts, are implicated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service