Skip to Content
Merck
All Photos(1)

Key Documents

700101P

Avanti

Dihydro FF-MAS-d6

Avanti Research - A Croda Brand

Synonym(s):

4,4-dimethyl(d6)-cholest-8(9), 14-dien-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,650.00

$1,650.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$1,650.00

About This Item

Empirical Formula (Hill Notation):
C29H42D6O
CAS Number:
Molecular Weight:
418.73
UNSPSC Code:
41141804
NACRES:
NA.12

$1,650.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 1 mg (700101P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]12CCC([C@](CC[C@H](O)C3(C([2H])([2H])[2H])C([2H])([2H])[2H])(C)C3CC4)=C4C1=CC[C@]2([H])[C@]([H])(C)CCCC(C)C

InChI

1S/C29H48O/c1-19(2)9-8-10-20(3)22-12-13-23-21-11-14-25-27(4,5)26(30)16-18-29(25,7)24(21)15-17-28(22,23)6/h13,19-20,22,25-26,30H,8-12,14-18H2,1-7H3/t20-,22-,25?,26+,28-,29-/m1/s1/i4D3,5D3

InChI key

OGQJUYXFIOFTMA-RHNFIJQVSA-N

General description

4,4-dimethyl(d6)-cholest-8(9), 14-dien-3β-ol (Dihydro FF-MAS-d6) is a deuterated derivative of dihydro FF-MAS. Dihydro follicular fluid meiosis-activating sterol (FF-MAS) is a sterol.[1]

Biochem/physiol Actions

Dihydro follicular fluid meiosis-activating sterol (FF-MAS) is an intermediate of the Kandutsch-Russell (K-R) pathway.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700101P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flux analysis of cholesterol biosynthesis in vivo reveals multiple tissue and cell-type specific pathways
Mitsche M A, et al.
eLife, 4, e07999-e07999 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service