Skip to Content
Merck
All Photos(2)

Key Documents

700048P

Avanti

24(R/S),25-epoxycholesterol-d6

Avanti Research - A Croda Brand

Synonym(s):

26,26,26,27,27,27-hexadeutero-24,25-epoxycholesterol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,640.00

$1,640.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$1,640.00

About This Item

Empirical Formula (Hill Notation):
C27H38O2D6
CAS Number:
Molecular Weight:
406.67
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.25

$1,640.00


Check Cart for Availability

Request a Bulk Order

description

24(R/S),25-epoxycholesterol-d6

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700048P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

24(R/S),25-epoxycholesterol-d6 is a deuterated side-chain substituted oxysterol.

Application

24(R/S),25-epoxycholesterol-d6 has been used as a primary and deuterated surrogate sterol standard in the quantitative analysis of extracted sterol samples from cultured cells and tissues.[1]

Biochem/physiol Actions

24(S),25-epoxycholesterol maintains cholesterol homeostasis. It modulates hepatic cholesterol metabolism in vivo. 24(S),25-epoxycholesterol is synthesized in the mevalonate pathway. It inhibits sterol regulatory element-binding protein (SREBP) activation.[2]
Deuterated sterol plays a vital role in chemical and biochemical research. It is used as a mechanistic tracer to study the role of steroid hormones in the function and modulation of several physiological processes. Deuterated sterol is also used as reference standards for mass spectrometry. 24(R/S),25-epoxycholesterol-d6 is a deuterium labeled sterol.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (700048P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Site-specific synthesis and application of Deuterium-Labeled sterols
Muchalski H, et al.
Archive for Organic Chemistry (2017)
24 (S), 25-epoxycholesterol: a messenger for cholesterol homeostasis
Brown AJ
The International Journal of Biochemistry & Cell Biology, 41(4), 744-747 (2019)
Extraction and analysis of sterols in biological matrices by high performance liquid chromatography electrospray ionization mass spectrometry
McDonald J, et al.
Test, 432, 145-170 (2007)
Jeffrey G McDonald et al.
Methods in enzymology, 432, 145-170 (2007-10-24)
We describe the development of a high performance liquid chromatography mass spectrometry (HPLC-MS) method that allows the identification and quantitation of sterols in mammalian cells and tissues. Bulk lipids are extracted from biological samples by a modified Bligh/Dyer procedure in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service