Skip to Content
Merck
All Photos(2)

Key Documents

700022P

Avanti

7-keto-27-hydroxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

3β,27-dihydroxy-5-cholesten-7-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$523.00

$523.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$523.00

About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$523.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700022P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

7-keto-27-hydroxycholesterol interconversion to 7β,(25R)26-dihydroxycholesterol (7β,26-diHC) is catalyzed by hydroxysteroid 11-β dehydrogenase.[1] It is also formed by the oxidation of 7-oxocholesterol (7-OC) in the presence of enzyme by sterol 26-hydroxylase (CYP27A1).[1]

Application

7-keto-27-hydroxycholesterol has been used as an oxysterol compound to study its interaction with smoothened (SMO) protein[2], as a substrate to 11β-hydroxysteroid dehydrogenases (11β-HSDs) for kinetic measurement studies[3],

Biochem/physiol Actions

7-keto-27-hydroxycholesterol (7-OC) acts as an agonist for the smoothened (SMO) protein of Hedgehog (Hh) signalling pathway, which is vital for proper cell differentiation in embryonic tissue. It elicits strong affinity to SMO compared to 7β,(25R)26-dihydroxycholesterol (7β,26-diHC).[1] 7-OC is reduced to 7β,(25R)26-dihydroxycholesterol (7β,26-diHC) by reactive oxygen species (ROS).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700022P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William J Griffiths et al.
Biochemical Society transactions, 47(2), 517-526 (2019-04-03)
In the present study, we discuss the recent developments in oxysterol research. Exciting results have been reported relating to the involvement of oxysterols in the fields of neurodegenerative disease, especially in Huntington's disease, Parkinson's disease and Alzheimer's disease; in signalling
Katharina R Beck et al.
Journal of lipid research, 60(9), 1535-1546 (2019-07-06)
Oxysterols previously were considered intermediates of bile acid and steroid hormone biosynthetic pathways. However, recent research has emphasized the roles of oxysterols in essential physiologic processes and in various diseases. Despite these discoveries, the metabolic pathways leading to the different
Benjamin R Myers et al.
Developmental cell, 26(4), 346-357 (2013-08-21)
Hedgehog (Hh) signaling during development and in postembryonic tissues requires activation of the 7TM oncoprotein Smoothened (Smo) by mechanisms that may involve endogenous lipidic modulators. Exogenous Smo ligands previously identified include the plant sterol cyclopamine (and its therapeutically useful synthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service