Skip to Content
Merck
All Photos(2)

Key Documents

700014P

Avanti

5β,6β-epoxycholestanol-d7

Avanti Research - A Croda Brand

Synonym(s):

111115

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$769.00

$769.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$769.00

About This Item

Empirical Formula (Hill Notation):
C27H39O2D7
CAS Number:
Molecular Weight:
409.70
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

$769.00


Check Cart for Availability

Request a Bulk Order

description

cholestanol, 5β,6β-epoxy-d7

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700014P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

5β,6β-epoxycholestanol-d7 is the deuterated version of 5β,6β-epoxycholestanol. Cholestanol is a metabolite of cholesterol.[1]

Application

5β,6β-epoxycholestanol-d7 has been used as an internal standard in isotope dilution-mass spectrometry analysis to quantify sterol and oxysterol from mitochondrial samples.[2] It has also been used as an oxysterol standard in high-performance liquid chromatography-mass spectrometry (HPLC-MS method).[3]

Biochem/physiol Actions

Cholestanol levels are elevated in biliary cirrhosis and in cholestasis.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700014P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentin Mutemberezi et al.
Journal of neuroinflammation, 15(1), 74-74 (2018-03-11)
Oxysterols are cholesterol derivatives that have been suggested to play a role in inflammatory diseases such as obesity, atherosclerosis, or neuroinflammatory diseases. However, the effect of neuroinflammation on oxysterol levels has only been partially studied so far. We used an
Julien Musman et al.
Biochemical pharmacology, 142, 87-95 (2017-06-25)
A major cause of cell death during myocardial ischemia-reperfusion is mitochondrial dysfunction. We previously showed that the reperfusion of an ischemic myocardium was associated with an accumulation of cholesterol into mitochondria and a concomitant strong generation of auto-oxidized oxysterols. The
Annika Mutanen et al.
The American journal of clinical nutrition, 100(4), 1085-1094 (2014-08-08)
Increased serum concentrations of plant sterols, including stigmasterol, during parenteral nutrition (PN) have been linked with serum biochemical signs of intestinal failure-associated liver disease (IFALD), whereas clinical data on their correlation to histologic liver injury have been limited. We studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service