Skip to Content
Merck
All Photos(2)

Key Documents

700009P

Avanti

15-ketocholestane

Avanti Research - A Croda Brand

Synonym(s):

3β-hydroxy-5α-cholestane-15-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$405.00

$405.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$405.00

About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$405.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700009P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

15-ketocholestane is the oxidized derivative of cholesterol[1] and is nonpolar lipid.[2]

Application

15-ketocholestane has been used as small molecule lipid to generate RAb2E9 recombinant monoclonal (RAb) using phage-library based screening).[3]

Biochem/physiol Actions

15-ketocholestane is a potential biomarker in multiple sclerosis and Autoimmune Encephalomyelitis (EAE).[3]

Packaging

5 mL Amber Glass Screw Cap Vial (700009P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel phage display-derived mycolic acid-specific antibodies with potential for tuberculosis diagnosis
Chan CE, et al.
Journal of Lipid Research, 54(10), 2924-2932 (2013)
Antigen microarrays identify unique serum autoantibody signatures in clinical and pathologic subtypes of multiple sclerosis
Quintana FJ, et al.
Proceedings of the National Academy of Sciences of the USA, 105(48), 18889-18894 (2008)
Generation and characterization of a novel recombinant antibody against 15-ketocholestane isolated by phage-display
Islam MD, et al.
International Journal of Molecular Sciences, 13(4), 4937-4948 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service