Skip to Content
Merck
All Photos(1)

Key Documents

699851P

Avanti

3A-MPLA

Avanti Research - A Croda Brand 699851P, powder

Synonym(s):

Monophosphoryl Tri-acyl Lipid A (Synthetic)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$5,620.00

$5,620.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$5,620.00

About This Item

Empirical Formula (Hill Notation):
C54H106N3O17P
CAS Number:
Molecular Weight:
1100.40
UNSPSC Code:
12352211
NACRES:
NA.25

$5,620.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (HPLC)

form

powder

packaging

pkg of 1 × 1 mg (699851P-1mg)
pkg of 1 × 5 mg (699851P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 699851P

application(s)

vaccine development

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]([C@@H](CO[C@@H]([C@@H]1NC(C[C@H](OC(CCCCCCCCCCCCC)=O)CCCCCCCCCCC)=O)O[C@H](CO)[C@@H](OP([O-])(O)=O)[C@@H]1O)O[C@H](O)[C@@H]2NC(C[C@H](O)CCCCCCCCCCC)=O)[C@@H]2O.[NH4+]

InChI

1S/C54H103N2O17P.H3N/c1-4-7-10-13-16-19-20-23-26-29-32-35-46(61)70-41(34-31-28-25-22-18-15-12-9-6-3)37-45(60)56-48-51(64)52(73-74(66,67)68)42(38-57)72-54(48)69-39-43-49(62)50(63)47(53(65)71-43)55-44(59)36-40(58)33-30-27-24-21-17-14-11-8-5-2;/h40-43,47-54,

InChI key

WBMOMCHVNLUEBA-MWYCKLIASA-N

General description

Monophosphoryl lipid A (MPLA) is an analog of the lipid A moiety.[1] It is regarded as a detoxified derivative of the lipopolysaccharide (LPS).[2]

Biochem/physiol Actions

Monophosphoryl lipid A (MPLA) is a natural agonist for the toll-like receptor-4 (TLR4). It is useful as an adjuvant in immunization. MPLA is a safe prophylactic agent and has immunotherapeutic applications.[3] It induces tumor necrosis factor and interleukin (IL)-1β, however not as efficient as lipopolysaccharide (LPS). MPLA improves immune response during bacterial sepsis infection.

Packaging

2 mL Amber Glass Crimp Cap Vial (699851P-1mg)
2 mL Amber Glass Crimp Cap Vial (699851P-5mg)

Other Notes

For R&D use only. Not for drug, household, or other uses.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The TLR4 agonist monophosphoryl lipid a drives broad resistance to infection via dynamic reprogramming of macrophage metabolism
Fensterheim BA, et al.
Journal of Immunology, 200(11), 3777-3789 (2018)
Monophosphoryl lipid A induces protection against LPS in medullary thick ascending limb through a TLR4-TRIF-PI3K signaling pathway
Watts III BA, et al.
American Journal of Physiology: Renal Physiology, 313(1), F103-F115 (2017)
Specific features of human monocytes activation by monophosphoryl lipid A
Chentouh R, et al.
Scientific reports, 8(1), 1-13 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service