Skip to Content
Merck
All Photos(1)

Documents

640014P

Avanti

Ceramide Synthase (CerS5/CerS6) Assay Kit

Avanti Research - A Croda Brand

Synonym(s):

Ceramide Synthase (CerS5/CerS6) Assay Kit, CerS5/S6 Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503

form

powder

usage

sufficient for ~275 reactions

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

The CerS Assay Kit provides an easy to use assay for measuring CerS activity in biological samples. This Kit offers the advantage of a short assay time requiring only small amounts of biological material while eliminating TLC as a separation technique with its attendant degradation of NBD sphinganine.

Biochem/physiol Actions

Ceramide synthase (cerS) catalyzes the acylation of sphinganine (dihydrosphingosine) to yield dihydroceramide and sphingosine to yield ceramide. There are six cerS (cerS1-6), which differ in their specificity to yield ceramides of different chain length. Ceramide is a key intermediate in sphingolipid metabolism and an key signaling molecule.

Packaging

Kit contains:
0.3mg 16:0 fatty acyl-CoA
0.1mg NBD Sphinganine

Storage and Stability

Note: this product should be stored at -20° for up to 6 months.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 810206PNBD Sphinganine, omega(7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R)-2-aminooctadecane-1,3-diol, powderSDS

  • 870716P16:0 Coenzyme A, Avanti Research - A Croda Brand 870716P, powderSDS

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The equilibrium between long and very long chain ceramides is important for the fate of the cell and can be influenced by co-expression of CerS.
Hartmann D, et al.
The International Journal of Biochemistry & Cell Biology, 45(7), 1195-1203 (2013)
Mammalian Ceramide Synthases
Michal L, et al.
IUBMB Life, 62(5), 347?356-347?356 (2010)
When do Lasses (longevity assurance genes) become CerS (ceramide synthases)? Insights into the regulation of ceramide synthesis.
Pewzner-Jung Y, et al.
The Journal of Biological Chemistry, 281(35), 25001-25005 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service