Skip to Content
Merck
All Photos(1)

Key Documents

330727P

Avanti

C15 Lactosyl(β) Ceramide-d7 (d18:1-D7/15:0)

Avanti Research - A Croda Brand

Synonym(s):

D-lactosyl-β-1,1′-N-pentadecanoyl-D-erythro-sphingosine-d7

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,430.00

$1,430.00


Available to ship on08 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 MG
$1,430.00

About This Item

Empirical Formula (Hill Notation):
C45H78D7NO13
CAS Number:
Molecular Weight:
855.20
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

$1,430.00


Available to ship on08 April 2025Details


Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (330727P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)[C@@]([H])(NC(CCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

InChI

1S/C45H85NO13/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-34(49)33(46-37(50)29-27-25-23-21-19-16-14-12-10-8-6-4-2)32-56-44-42(55)40(53)43(36(31-48)58-44)59-45-41(54)39(52)38(51)35(30-47)57-45/h26,28,33-36,38-45,47-49,51-55H,3-25,27,29-32H2,1-2H3,(H,46,50)/b2

InChI key

WCBBCGOUIWFKOI-LBQLNCQHSA-N

General description

C15 Lactosyl(β) Ceramide-d7 (d18:1-D7/15:0) is a deuterated derivative of C15 Lactosyl(β) Ceramide. Lactosyl ceramide is a cerebroside, that contains a lactose moiety (disaccharide) linked β-glycosidically to the 1-position carbon.[1]

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dietary and endogenous sphingolipid metabolism in chronic inflammation
Norris G H, and Blesso C N
Nutrients, 9(11), 1180-1180 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service