Skip to Content
Merck
All Photos(1)

Key Documents

330717C

Avanti

15:0-18:1 PE

Avanti Research - A Croda Brand

Synonym(s):

1-pentadecanoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$370.00

$370.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$370.00

About This Item

Empirical Formula (Hill Notation):
C38H74NO8P
CAS Number:
Molecular Weight:
703.97
UNSPSC Code:
51191904
NACRES:
NA.25

$370.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

chloroform solution

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[NH3+])(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCC)=O)=O

InChI key

ADCNXGARWPJRBV-UVCQAILXSA-N

Packaging

2 mL Amber Glass Sealed Ampule (330717C-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service