Skip to Content
Merck
All Photos(1)

Key Documents

330612

Avanti

DOPC ELISA (Clear)

Avanti Research - A Croda Brand

Synonym(s):

DOPC ELISA, DOPC ELISA Snoopers, Clear, for use in absorbance applications

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$62.80

$62.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$62.80

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

$62.80


Check Cart for Availability

Request a Bulk Order

form

liquid

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

1 ea of 1 × (ELISA strip packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

General description

ELISA Snoopers are 8-well strips pre-coated with a single high-quality lipid species. They can be used to investigate protein-lipid interactions, and characterize and titrate lipid binding proteins. This format provides the following advantages:
  • quantitative lipid-protein binding studies
  • assemble a 96-well plate using only the lipids of interest for binding studies
  • flexibility - use 8 to 96 wells, and 1 to 12 lipids as needed.

The 8-well strips are available in optically clear, white, or black surfaces for use in colormetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.

Packaging

ELISA Strip Packaged in Foil Pouch (330612-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service