Skip to Content
Merck
All Photos(1)

Key Documents

330256W

Avanti

Cholic acid-d4

Avanti Research - A Croda Brand

Synonym(s):

5ß-cholanic acid-3a,7a,12a-triol (2,2,4,4-d4)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$250.00

$250.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$250.00

About This Item

Empirical Formula (Hill Notation):
C24H36D4O5
CAS Number:
Molecular Weight:
412.60
UNSPSC Code:
41141802
NACRES:
NA.12

$250.00


Check Cart for Availability

Request a Bulk Order

form

methanol solution

packaging

package of 1 × 100 μg

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

100 μg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC([2H])([2H])[C@@H](O)C3([2H])[2H])(C)[C@@]3([H])C[C@H]2O)([H])C[C@H](O)[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@@]([H])(C)CCC(O)=O

InChI

1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15+,16+,17-,18-,19+,20-,22-,23-,24+/m0/s1/i8D2,10D2

InChI key

BHQCQFFYRZLCQQ-ONSXQPQQSA-N

Packaging

2 mL Amber Glass Sealed Ampule (330279W-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service