Skip to Content
Merck
All Photos(1)

Documents

131101C

Avanti

Brain Extract Total

Avanti Research - A Croda Brand

Synonym(s):

Brain Total Lipid Extract (Porcine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

chloroform solution

packaging

pkg of 2 × 20 mL (131101C-1g)
pkg of 1 × 4 mL (131101C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (131101C-100mg)
25 mg/mL (131101C-1g)

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Total Lipid Extract is a chloroform:methanol extract of the respective tissue (Brain, Soybean, Heart, etc.). This extract is partitioned against deionized water, and the chloroform phase is concentrated.

Application

Brain extract total has been used as a lipid carrier for the extraction and deacetylation of lipids.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (131101C-1g)
5 mL Clear Glass Sealed Ampule (131101C-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziyi Yang et al.
Bioorganic & medicinal chemistry, 28(7), 115374-115374 (2020-02-25)
A series of novel flurbiprofen-clioquinol hybrids were designed and synthesized as multifunctional agents for Alzheimer's disease therapy, and their potential was evaluated through various biological experiments. In vitro studies showed that most target compounds exhibited significant ability to inhibit self-
Phosphoinositide Analysis using the HPLC System Equipped with a Fraction Collector and the TSKgel SAX Column
KITA S, et al.
Medical bulletin of Fukuoka University, 42(1), 175-181 (2015)
Emmanuel Boucrot et al.
Cell, 149(1), 124-136 (2012-04-03)
Shallow hydrophobic insertions and crescent-shaped BAR scaffolds promote membrane curvature. Here, we investigate membrane fission by shallow hydrophobic insertions quantitatively and mechanistically. We provide evidence that membrane insertion of the ENTH domain of epsin leads to liposome vesiculation, and that
Wei Li et al.
Bioorganic chemistry, 97, 103707-103707 (2020-03-09)
A series of pyridoxine-resveratrol hybrids were designed and synthesized as monoamine oxidase B inhibitors for the treatment of Parkinson's disease. Most of them exhibited potent inhibitory activities on MAO-B with high selectivity. Specifically, compounds 12a, 12g and 12l showed the
B Sandrino et al.
Chemistry and physics of lipids, 230, 104930-104930 (2020-05-30)
Understanding protein aggregation is essential to unveil molecular mechanisms associated with neurodegenerative diseases such as Alzheimer's, Huntington's and spongiform encephalopathy, particularly to determine the role of interaction with cell membranes. In this study, we employ Langmuir monolayers as cell membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service