ATEH998DEA8C
AstaTech, Inc.
4-AMINOTHIOMORPHOLINE 1,1-DIOXIDE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-thiomorpholinamine 1,1-dioxide
Inchi Code
1S/C4H10N2O2S/c5-6-1-3-9(7,8)4-2-6/h1-5H2
InChI key
YDKNASMIEZGJEJ-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Physical Form
Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=159821
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=T70689
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service