Skip to Content
Merck
All Photos(1)

Key Documents

ATEH93EDCD0F

AstaTech, Inc.

1-BENZYL 4-(TERT-BUTYL) (2R,5R)-2-(HYDROXYMETHYL)-5-METHYLPIPERAZINE-1,4-DICARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
364.44
MDL number:

Iupac Name

1-benzyl 4-(tert-butyl) (2R,5R)-2-(hydroxymethyl)-5-methylpiperazine-1,4-dicarboxylate

Inchi Code

1S/C19H28N2O5/c1-14-10-21(17(23)25-13-15-8-6-5-7-9-15)16(12-22)11-20(14)18(24)26-19(2,3)4/h5-9,14,16,22H,10-13H2,1-4H3/t14-,16-/m1/s1

InChI key

DVPLXTZRAMPJQO-GDBMZVCRSA-N

Purity

95%

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=174707

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=C92269


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library