Skip to Content
Merck
All Photos(1)

Key Documents

ATE429354799

AstaTech, Inc.

TERT-BUTYL 4-(4-AMINOPYRIMIDIN-2-YL)PIPERAZINE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
279.34
MDL number:
Pricing and availability is not currently available.

Iupac Name

tert-butyl 4-(4-amino-2-pyrimidinyl)-1-piperazinecarboxylate

Inchi Code

1S/C13H21N5O2/c1-13(2,3)20-12(19)18-8-6-17(7-9-18)11-15-5-4-10(14)16-11/h4-5H,6-9H2,1-3H3,(H2,14,15,16)

InChI key

KVQHCKUTCGHMBO-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=45378

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=CS1362


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library