Skip to Content
Merck
All Photos(1)

Key Documents

ATE386054369

AstaTech, Inc.

HEXAHYDRO-1H-PYRIDO[1,2-A]PYRAZIN-4(6H)-ONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
154.21
MDL number:

Iupac Name

octahydro-4H-pyrido[1,2-a]pyrazin-4-one

Inchi Code

1S/C8H14N2O/c11-8-6-9-5-7-3-1-2-4-10(7)8/h7,9H,1-6H2

InChI key

DPTQSVONRGAROK-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=14445

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=68528


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library