Skip to Content
Merck
All Photos(1)

Key Documents

APOH316CDCDC

Apollo Scientific Ltd

3-Iodo-N,N-dimethylbenzamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
275.09
MDL number:

Iupac Name

3-iodo-N,N-dimethylbenzamide

Inchi Code

1S/C9H10INO/c1-11(2)9(12)7-4-3-5-8(10)6-7/h3-6H,1-2H3

InChI key

XSLDOPYONMGECY-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/3-iodo-nn-dimethylbenzamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-iodo-nn-dimethylbenzamide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library