APO455831626
Apollo Scientific Ltd
2-Ethynyl-5-methoxypyrazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-ethynyl-5-methoxypyrazine
Inchi Code
1S/C7H6N2O/c1-3-6-4-9-7(10-2)5-8-6/h1,4-5H,2H3
InChI key
CGBDPVUIUFCVQS-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/2-ethynyl-5-methoxypyrazine
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/2-ethynyl-5-methoxypyrazine
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.