Skip to Content
Merck
All Photos(1)

Key Documents

APO455825722

Apollo Scientific Ltd

2-[4-(1,3-Dithiolan-2-yl)phenoxy]-N'-hydroxyethanimidamide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
270.38
MDL number:

bp

113-115

Iupac Name

(1Z)-2-[4-(1,3-dithiolan-2-yl)phenoxy]-N'-hydroxyethanimidamide

Inchi Code

1S/C11H14N2O2S2/c12-10(13-14)7-15-9-3-1-8(2-4-9)11-16-5-6-17-11/h1-4,10-11H,5-7,12H2

InChI key

OUPLHVABVMUCDV-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/2-4-13-dithiolan-2-ylphenoxy-n-hydroxyethanimidamide

Boiling point

113-115

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-4-13-dithiolan-2-ylphenoxy-n-hydroxyethanimidamide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library