APO428498596
Apollo Scientific Ltd
4-Ethoxy-3-(trifluoromethyl)phenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-ethoxy-3-(trifluoromethyl)phenol
Inchi Code
1S/C9H9F3O2/c1-2-14-8-4-3-6(13)5-7(8)9(10,11)12/h3-5,13H,2H2,1H3
InChI key
KTAPXNQLDCIIAO-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
98%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Liquid
Product Link
https://store.apolloscientific.co.uk/product/4-ethoxy-3-trifluoromethylphenol
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/4-ethoxy-3-trifluoromethylphenol
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service