AMBH9614FDB4
Ambeed, Inc.
4-Phenyl-1,2,3,4-tetrahydroquinoline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-phenyl-1,2,3,4-tetrahydroquinoline
Inchi Code
1S/C15H15N/c1-2-6-12(7-3-1)13-10-11-16-15-9-5-4-8-14(13)15/h1-9,13,16H,10-11H2
InChI key
YUEVNAMKBJHWDG-UHFFFAOYSA-N
storage temp.
Keep in dark place,Sealed in dry,Room Temperature
Purity
95%
Country of Origin
CN
Shipping Temperature
Normal
Physical Form
solid
Product Link
https://www.ambeed.com/products/30290-78-9.html
Synonym
4-phenyl-1,2,3,4-tetrahydroquinoline
Signal Word
Warning
Hazard Statements
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.