Skip to Content
Merck

AMBH2D6F813C

Ambeed, Inc.

Quinuclidine-4-carboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$448.92

$448.92

List Price$522.00Save 14%

Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB04901


Select a Size

Change View
100 μG
$448.92

About This Item

CAS Number:
Molecular Weight:
191.66
MDL number:

$448.92

List Price$522.00Save 14%

Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB04901

Iupac Name

quinuclidine-4-carboxylic acid hydrochloride

Inchi Code

1S/C8H13NO2.ClH/c10-7(11)8-1-4-9(5-2-8)6-3-8;/h1-6H2,(H,10,11);1H

InChI key

JTYXRFSULOZNPH-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB5200988SAB5200888SAB5200904
conjugate

unconjugated

conjugate

Allophycocyanin conjugate

conjugate

unconjugated

conjugate

phycoerythrin (R-PE) conjugate

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

mouse, human, rat

species reactivity

human, mouse, rat

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

Gene Information

rat ... Grin2a(24409)

clone

S327A-38, monoclonal

clone

S327A-38, monoclonal

clone

S327-95, monoclonal

clone

S327-95, monoclonal

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

storage temp.

Inert atmosphere,Room Temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/40117-63-3.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.ambeed.com/products/40117-63-3.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service