Skip to Content
Merck
All Photos(1)

Key Documents

Z765147

Tygon® formula E-3603 laboratory tubing

I.D. × O.D. × wall thickness 1/2 in. × 5/8 in. × 1/16 in.

Synonym(s):

DEHP tubing alternative, chemical resistant tubing, laboratory tubing, vacuum tubing

Sign Into View Organizational & Contract Pricing

Select a Size

50 FT
$172.00

$172.00


Available to ship on14 April 2025Details



Select a Size

Change View
50 FT
$172.00

About This Item

UNSPSC Code:
41105108
NACRES:
NB.17

$172.00


Available to ship on14 April 2025Details


packaging

pkg of 50 feet

manufacturer/tradename

Tygon

I.D. × O.D. × wall thickness

1/2 in. × 5/8 in. × 1/16 in.

Looking for similar products? Visit Product Comparison Guide

General description

TYGON® E-3603 non-DEHP laboratory and vacuum tubing delivers the same superior performance you′ve come to expect from TYGON products - but now in a formulation that contains a widely accepted substitute for DEHP plasticizer.

TYGON® E-3603 laboratory tubing is specially formulated for resistance to flex fatigue and abrasion, handles virtually all inorganic chemicals found in the lab, and is less permeable than rubber tubing. The glassy-smooth inner bore helps prevent build-up to facilitate cleaning.

TYGON® E-3603 vacuum tubing shares all these performance features and has extra heavy walls that will withstand a full vacuum at room temperature.

Features/Benefits:
  • Alternative to DEHP tubing
  • New formulation delivers the same excellent performance
  • Contains no BPA
  • Outstanding chemical resistance
  • Lot-to-lot consistency for reproducible results
  • Increases productivity in peristaltic pumps - outlasts other clear tubing 2 to 1
  • Smooth, polished inner bore
  • Non-oxidizing and non-contaminating
  • Slips easily over fittings and grips securely
  • Coils marked at 1-foot intervals for easy measuring
  • Meets FDA criteria

Typical Applications:
  • General laboratory
  • Analytical instruments
  • Peristaltic and vacuum pumps
  • Ideal for condensers, incubators, desiccators, gas lines and drain lines

Legal Information

Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service