Key Documents
Z758973
Bollé Atom safety goggles
ANSI compliant
Synonym(s):
protective eyewear, protective glasses, safety eyeglasses, safety eyewear, safety glasses, safety spectacles
Select a Size
1 EA
$119.00
$119.00
Check Cart for Availability
Select a Size
Change View
1 EA
$119.00
About This Item
UNSPSC Code:
46181802
$119.00
Check Cart for Availability
feature
ANSI compliant
packaging
pkg of 1 ea
manufacturer/tradename
Bollé 40092
General description
These Bollé Atom safety goggles are lightweight and compact and fit over prescription spectacles as well as with a respiratory half mask. There is an adjustable strap and a liquid overflow chute. The goggles are Thermo Plastic Rubber (TPR) ventilated.
  • Anti-scratch, and anti-fog coatings
  • Adjustable strap
  • Ball-pivot
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)