Skip to Content
Merck
All Photos(1)

Key Documents

Z743911

ART® SoftFit L Reload System empty reload HLT insert

for 1000 ul - 1200 ul racks

Synonym(s):

Reload rack, tip rack

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 8 EA
$533.00

$533.00


Check Cart for Availability


Select a Size

Change View
4 X 8 EA
$533.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
feature:
for 1000 ul - 1200 ul racks
packaging:
pkg of 4X8 ea
manufacturer/tradename:
Molecular BioProducts 3706-ER

$533.00


Check Cart for Availability

feature

for 1000 ul - 1200 ul racks

packaging

pkg of 4X8 ea

manufacturer/tradename

Molecular BioProducts 3706-ER

technique(s)

PCR: suitable

Looking for similar products? Visit Product Comparison Guide

General description

The SoftFit-L empty racks provide an eco-friendly alternative for packaging. They can be paired with SoftFit-L reload inserts and reload towers to eliminate excessive plastic waste that is typically produced with conventional racks. These racks are specifically made to be compatible with ART SoftFit-L reload inserts and reload towers.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service