Key Documents
Z743250
Julabo® bath spring clamps for Erlenmeyer flasks
fits 300 mL flask
Select a Size
1 EA
$107.00
$107.00
Estimated to ship on25 April 2025
Select a Size
Change View
1 EA
$107.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.77
$107.00
Estimated to ship on25 April 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970607
General description
The individual spring clips can be positioned in the shaking water bath using the base tray for spring clips (Z743201-1EA). A maximum of 15 spring clips for 300 ml Erlenmeyer flasks can be mounted on a base tray.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)