Key Documents
Z743167
Julabo® bath covers
flat cover to fit CORIO CD-B39
Select a Size
1 EA
$606.00
$606.00
Estimated to ship on05 April 2025
Select a Size
Change View
1 EA
$606.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
$606.00
Estimated to ship on05 April 2025
packaging
pack of 1 ea
manufacturer/tradename
Julabo 9970293
General description
Flat stainless steel bath cover reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)