Key Documents
Z742514
Block for digital dry bath and MyBlock
solid block for slides or custom machining
Select a Size
1 EA
$206.00
$206.00
Check Cart for Availability
Select a Size
Change View
1 EA
$206.00
About This Item
UNSPSC Code:
41102404
NACRES:
NB.77
$206.00
Check Cart for Availability
feature
solid block for slides or custom machining
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific BSW01
General description
For use with Benchmark Digital Dry Bath
  • Material: high grade aluminum
  • Precision machined to match the tube shape and size, providing unsurpassed temperature uniformity
Legal Information
MyBlock is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)