Key Documents
Z742304
Tube holder for RotoBot Rotating Mixers
holds 1 x 15 mL tubes, pack of 2 ea
Select a Size
1 EA
$34.10
$34.10
Check Cart for Availability
Select a Size
Change View
1 EA
$34.10
About This Item
UNSPSC Code:
41103811
NACRES:
NB.61
$34.10
Check Cart for Availability
feature
holds 1 x 15 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Benchmark Scientific R4040-150
table diam.
16-17 mm
General description
For higher capacity applications, these holders can be purchased as accessories for the Rotobot and Rotobot Mini mixers. Horizontal holders are available for applications such as Western blotting and hybridization. The unique, sliding attachment allows for quick and easy changing of tube holders.
Legal Information
Rotobot is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)