Z740148
XPell™ R indicating reagent powder for peroxides prevention
pkg of 1 ea
Select a Size
$243.00
Select a Size
About This Item
$243.00
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Xplosafe, LLC 3004
General description
XPell R features:
- Safely neutralizes peroxide build up for less expensive disposal
- Visual solution - distinct blue color indicates the absence of peroxides
- Simple and effective, does not require any additional materials or steps
- No special handling, storage or disposal requirements
- Significant cost savings by eliminating peroxide hazard and disposal
- Improved laboratory safety and prevention of peroxide related explosions/accidents
Directions:
Before disposal, add approximately 0.5 g of XPell R powder for each liter of solvent. XPell R neutralizes peroxides present in the solvent and peroxides that form over time. The solvent should be free of peroxides as long as the immersed powder remains dark blue. A color change to a yellow/pale color indicates that the protection is exhausted. If this change occurs, add more powder to the solvent until the color of the additional powder remains dark blue.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service