Skip to Content
Merck
All Photos(1)

Key Documents

Z699381

Swabs and SwabChecks

Total Count Swab Kit

Synonym(s):

swab kit, swab kits, swabs

Sign Into View Organizational & Contract Pricing

Select a Size

30 EA
$385.00

$385.00


Check Cart for Availability


Select a Size

Change View
30 EA
$385.00

About This Item

UNSPSC Code:
41111700
NACRES:
NB.79

$385.00


Check Cart for Availability

feature

Total Count Swab Kit

packaging

pkg of 30 ea

manufacturer/tradename

Maine Mfg 10498315

application(s)

environmental
food and beverages

suitability

bacteria

Looking for similar products? Visit Product Comparison Guide

General description

The SwabCheck principle
The surface is wiped with a cellulose swab and any bacteria collected are transferred via the swab into a tube containing a special medium with an indicator dye, which is then incubated. A single bacterium is sufficient to cause a color change. This means that SwabCheck is about 1000 times more sensitive than the conventional ATP method. This accuracy is particularly important in the food industry. With this simple method, it is possible to identify microorganisms such as Listeria monocytogenes, which must not be present in any concentration in food and beverages.

Features and Benefits
  • The right test for each type of contamination
  • Qualitative and semi-quantitative hygiene control
  • Sterile packed and ready-for-use
  • Easy to handle
  • Rapid results
  • Long shelf-life
  • SwabCheck use

Handling is easy. Open the sterile pack, remove the swab and wipe it over an area of about 10 x 10 cm. Then twist off the cap of the medium tube and insert the swab so that the cap fits tightly. Label the sample tube and incubate at the appropriate temperature.

A change in color indicates the presence of the microorganism in question. The quicker the color change occurs, the higher the bioburden. If no color change has been observed after the maximum incubation period has elapsed, then the corresponding microorganism is not present.GVS offers SwabCheck in packs of 25 pieces. Shelf-life of 12 months.
Total Count Swab Kit
Used for the non-selective development and enumeration of all aerobic bacteria on surfaces in accordance with Hazard Analysis and Critical Control Points (HACCP). The kit includes the swabs and culture medium, packaged with a membrane device, providing a quantitative result. All bacteria develop on TGE media and produce a range of different colored and sized colonies. It is not possible using TGE to presumptively identify any bacteria. Identification can only be undertaken using traditional microbiology techniques following initial colony development.

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are the swab and swabchecks good for testing for Ecoli and HPC? aldreich Z699381

    1 answer
    1. This product does not have specificity. It is used for the non-selective development and enumeration of all aerobic bacteria on surfaces. It is not possible using TGE to presumptively identify any bacteria. Identification can only be undertaken using traditional microbiology techniques following initial colony development.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service