Key Documents
Z690503
Ace all-PTFE filter reactor assembly
capacity 1 L, joint: ST/NS 24/40
Select a Size
1 EA
$14,940.00
$14,940.00
Check Cart for Availability
Select a Size
Change View
1 EA
$14,940.00
About This Item
UNSPSC Code:
41103817
NACRES:
NB.51
$14,940.00
Check Cart for Availability
Joint
joint: ST/NS24/40
manufacturer/tradename
Ace Glass 6390-10
capacity
1 L
Related Categories
General description
These PTFE filter reactors can be used for fluorine chemistry and with strong alkaline compounds that can etch glass. The head, head joints, body, stirrer-bearing, bottom filter assembly with valve, and stirring shaft with agitator are all PTFE. Even the O-Rings are PTFE encapsulated. The filter supports are polypropylene. The bottom filter assembly is either #50 or #80 Ace-Thred and threads in/out for easy capture of filtrate, or for cleaning. A Halar® coated stainless steel internal coil is included (in the 1 L size) for heating or cooling the contents. IKA RW 20 over head stirrer recommended.
Legal Information
Halar is a registered trademark of Solvay Group
Choose from one of the most recent versions:
Certificates of Analysis (COA)