Key Documents
Z682071
Ace cold trap outer body only for condenser "C" assembly
plastic-coated glass, fits Büchi® models R-200/R-205 and R-114/R-144
Select a Size
1 EA
$1,710.00
$1,710.00
Check Cart for Availability
Select a Size
Change View
1 EA
$1,710.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$1,710.00
Check Cart for Availability
material
plastic-coated glass
manufacturer/tradename
Büchi equivalent, 40643
Related Categories
General description
The cold trap outer body for the Ace condenser "C" assembly, is glass with a 35/20 inner spherical joint bottom connection that attaches to the receiver flasks. Features standard top and side flange that connects directly to the Büchi rotary evaporator.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)