Key Documents
Z677876
Finntip® Flex pipette tips
Finntip Flex 200, sterile
Select a Size
960 EA
$252.00
$252.00
Available to ship on22 April 2025Details
Select a Size
Change View
960 EA
$252.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
sterility:
sterile
feature:
barrier: no
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Finn 94060313
$252.00
Available to ship on22 April 2025Details
material
colorless tip
yellow
sterility
sterile
feature
barrier: no
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Finn 94060313
maximum volume
200 μL
volume range
1-200 μL
General description
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Other Notes
Browse all pipette tips
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)