Skip to Content
Merck
All Photos(1)

Documents

Z677701

Vitoject fluorocarbon rubber gloves

size M (9)

Synonym(s):

chemically resistant gloves, hand protection against aggressive chemicals

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
46181504
NACRES:
NB.35

material

black (fluorocarbon rubber)
black gloves

size

M (9)

sterility

non-sterile

feature

CE compliant
latex free
powder-free: no
reusable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)

packaging

pack of 1 pair

manufacturer/tradename

KCL 890

L × thickness

350 mm × 0.7 mm

Looking for similar products? Visit Product Comparison Guide

General description

Vitoject is made of 100% high chemical resistant Viton®, absolutely indispensable when working with aromatic hydrocarbons (Benzene, Toluene, ...), chlorinated hydrocarbons (Tri, Per, ...), fuels, aggressive acids and many other chemicals.

Manufactured with the ecologically beneficial and modern Vulcoject-process technology (injection molding).

  • Very good protection against extremely aggressive chemicals
  • 100% gastight
  • Excellent wearing comfort
  • Solvent free by eco-friendly injection molding process
  • CE compliant

Legal Information

Vitoject is a trademark of KCL GmbH
Viton is a registered trademark of E. I. du Pont de Nemours and Company

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service