Key Documents
Z645214
IKA® M 21 spare cutter
M 21 spare cutter, stainless steel
Select a Size
1 EA
$315.00
$315.00
Check Cart for Availability
Select a Size
Change View
1 EA
$315.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
$315.00
Check Cart for Availability
material
stainless steel
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 328200
suitability
suitable for (crushing materials up to Mohs hardness 5)
General description
Stainless steel replacement bat for use with the IKA M 20 Universal Mill, for breaking substances up to Mohs hardness 5.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)