Merck
All Photos(4)
Documents
Z635340
Swiss Boy lab jacks
load ≤80 kg (middle position), platform L × W 122 mm × 150 mm, Model 115
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
manufacturer/tradename
Grauer
parameter
≤80 kg load (middle position)
80-278 mm vertical limit
platform L × W
122 mm × 150 mm
color
yellow
General description
Jack can be raised with load in place.
These high quality Swiss made jacks have massive cast aluminum frames with a chemically resistant anodised finish. Swiss boy jacks are designed to carry heavy loads safely. Available either in standard size or with extended carrying plates for large pots or pans. Units carrying more that 90kg are also available with a motorized lift adjustment for enhanced convenience. The max load is for the middle position. Supplied with top and bottom plates.
Products with motorized jacks are available in Europe only.
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2