Skip to Content
Merck
All Photos(1)

Key Documents

Z567477

NMR tube caps

NMR tube size 2 mm, red

Synonym(s):

cap tubes, nmr tubes, plastic end cap

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
$23.30

$23.30


Estimated to ship on30 March 2025



Select a Size

Change View
25 EA
$23.30

About This Item

UNSPSC Code:
41121800
NACRES:
SB.32

$23.30


Estimated to ship on30 March 2025


packaging

pkg of 25 ea

manufacturer/tradename

Wilmad 521-T

NMR tube size

2 mm

color

red

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service