Key Documents
Z556866
Sigma-Aldrich
Aldrich® thermometer extension Benchclamp
nickel plated (finish), pkg of 1 ea
Select a Size
1 EA
$71.10
$71.10
Check Cart for Availability
Select a Size
Change View
1 EA
$71.10
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
$71.10
Check Cart for Availability
material
nickel plated (finish)
packaging
pkg of 1 ea
L
210 mm
jaw capacity
6-12 mm
Related Categories
General description
Nickel-plated finish is corrosion resistant. Knurled thumbscrew thightens jaw to hold objects firmly.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)