Z552380
Aldrich® cold-finger condenser
joint: ST/NS 14/20
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS14/20
Overall H
180 mm
cold-finger L × diam.
180 mm × 54 mm
Related Categories
General description
For reactions using low-boiling solvents.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC