Z551171
Graduated Valumetric™ bottles
black phenolic cap (with Poly-Seal® liner), capacity 30 mL (1 oz)
Synonym(s):
W216976
Select a Size
$253.00
Select a Size
About This Item
$253.00
Recommended Products
material
black phenolic cap (with Poly-Seal® liner)
glass bottle
round bottle
Joint
Joints threaded neck (33-400)
feature
with cap
packaging
pkg of 48 ea
manufacturer/tradename
WHEATON® W216976
capacity
30 mL (1 oz)
diam. × H
34 mm × 68 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Additional graduation marks and an opposite scaling (accurate to ± 5 %) simplify the reading of the filling level as well as the quantity withdrawn
- Manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- Reliable sealing due to PTFE-coated cap liner
- TÜV tested thermal shock resistance of 160 K
- Complete with PFA pouring ring and cap
- Standardised GL 45 thread and matching GL 45 cap for a realiable seal
- Simple, drip free pouring with resistant PFA pouring ring
- Supplied complete with cap and pouring ring
- Colourant free materials of construction for lower extractables profile
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service