Key Documents
Z549185
Sigma-Aldrich
Aldrich® antisplash splash-guard adapter without return hole
female joint: ST/NS 29/32, male joint: ST/NS 24/40, capacity 500 mL
Select a Size
1 EA
$240.00
$240.00
Estimated to ship on12 April 2025
Select a Size
Change View
1 EA
$240.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$240.00
Estimated to ship on12 April 2025
Joint
female joint: ST/NS29/32
male joint: ST/NS24/40
capacity
500 mL
General description
Aldrich® antisplash splash-guard adapter without return hole are traps that greatly reduce carry-over from foaming or bumping. Whatever comes up into this trap will stay and not go back to the evaporator flask.
Application
Aldrich® antisplash splash-guard adapter without return hole is ideal for use between the vapor tube and rotary evaporator flask to prevent solvent from being drawn into condenser in case of "bumping".
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)