Z544493
Molymod® molecular model set
teacher/researcher set, organic stereochemistry
Select a Size
$204.00
Available to ship on28 April 2025Details
Select a Size
About This Item
$204.00
Available to ship on28 April 2025Details
Recommended Products
feature
no. of atoms 168
no. of bonds 192
manufacturer/tradename
Molymod MMS-050
General description
60 molydomes hydrogen
44 Carbon -black ( 2 different types)
1 group alkene
1 group alkyne
16 Oxygen - red ( 3 different types)
4 Hydrogen - white
12 Nitrogen - blue ( 2 different types)
12 Sulphur - yellow ( 2 different types)
5 Phosphorus - purple ( 2 different types)
1 Metal - grey
8 chlorine
2 bromine
2 iodine
6 p-lobes pink
6 p-lobes purple
40 links, gray, medium ML-12
12 links, gray, flexible long ML-13
60 short colorles
60 short white
20 V-links
2 link remover tool
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)