Key Documents
Z531286
Sigma-Aldrich
Aldrich® coiled condenser
jacketed, L 300 mm, joint: ST/NS 24/40
Select a Size
1 EA
$636.00
$636.00
Estimated to ship on20 April 2025
Select a Size
Change View
1 EA
$636.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$636.00
Estimated to ship on20 April 2025
Joint
joint: ST/NS24/40
L
300 mm
Related Categories
General description
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)