Skip to Content
Merck
All Photos(1)

Key Documents

Z528757

Sigma-Aldrich

Scienceware® Poxygrid® baskets

basket L × W × H 51 cm × 28 cm × 9 cm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$133.00

$133.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 EA
$133.00

About This Item

UNSPSC Code:
41103200
NACRES:
NB.35

$133.00


Estimated to ship on01 June 2025


packaging

pack of 1 ea

manufacturer/tradename

Bel-Art H16767-0000

basket L × W × H

51 cm × 28 cm × 9 cm

Looking for similar products? Visit Product Comparison Guide

General description

Epoxy-coated steel. Use in transport or batch dipping glassware.

Legal Information

Poxygrid is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service