Key Documents
Z514950
Kimax® high strength centrifuge tube replacement cap
for Z514896
Synonym(s):
73802-20400
Select a Size
1 PKG
$369.00
$369.00
Check Cart for Availability
Select a Size
Change View
1 PKG
$369.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
$369.00
Check Cart for Availability
manufacturer/tradename
Kimble®
Features and Benefits
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)