Key Documents
Z510793
Sigma-Aldrich
Connecting Adapter
female joint: ST/NS 19/22, thread size 13-425
Synonym(s):
747130-1319
Select a Size
1 EA
$70.20
$70.20
Available to ship on22 April 2025Details
Select a Size
Change View
1 EA
$70.20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$70.20
Available to ship on22 April 2025Details
thread size
13-425
Joint
female joint: ST/NS19/22
General description
Connects ground joint glassware to threaded microscale component or sample vials. Excellent for use with splash-guards to recover small quantities of material during rotary evaporation. Order threaded vial connectors separately.
Features and Benefits
  • Accessory for microscale MICROFLEX threaded kits
  • Connects Standard Taper outer ground joint to GPI thread of inner ground joint
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Choose from one of the most recent versions:
Certificates of Analysis (COA)